Order Entry
Denmark
ContactUsLinkComponent
In vivo-jetpei-fluof is a fluorescently labeled dna and sirna delivery reagent for in vivo experiments in animal models (fluorescein -conjugated, green label, excitation at 490 nm emission at 520 nm). 1 * 100 µl
Catalog # 205-10G
undefined
In vivo-jetpei-fluof is a fluorescently labeled dna and sirna delivery reagent for in vivo experiments in animal models (fluorescein -conjugated, green label, excitation at 490 nm emission at 520 nm). 1 * 100 µl
Catalog # 205-10G