Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

PolySat® Pre-Saturated Wipes

TX1051, 280×230 mm, 50 wipes/flexpack, 24 flexpacks/case
  115-0301
 :   Texwipe
PolySat® Pre-Saturated Wipes
  115-0301
 :  Texwipe
PolySat® Pre-Saturated Wipes
PolySat® Pre-Saturated Wipes

TX1051, 280×230 mm, 50 wipes/flexpack, 24 flexpacks/case

 
5.570,00 kr
   
1 * 1.200 Items
 

PolySat® is constructed from a 100% melt-blown polypropylene substrate that is pre-wetted with 70% isopropyl alcohol and packaged in a resealable FlexPack. PolySat® is convenient for cleaning sensitive surfaces and polishing equipment in controlled environments.

  • FlexPacks are a convenient alternative to wipes and spray bottles, eliminating mixing, sterilising and waste
  • Packed in SolventSafe plastic that can be wiped down prior to introduction into the cleanroom
  • Delivers a pre-measured amount of high purity alcohol to the surface: Optimises alcohol usage and reduces spill potential
  • Low particle generation and low levels of solvent extractables

Colour: White

Caution: The product(s) on this page are outside the scope of the EU Directive 2019/904.

 
  • Packed:
    50 wipes/flexpack, 24 flexpacks/case
  • Description:
    TX1051
  • L×W:
    280×230 mm
  • Seq:
    2
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.