Order Entry
Denmark
Orders LinkContactUsLinkComponent
Grounding cables, Super Erecta®
Grounding cables, Super Erecta®
Grounding cable with spring-loaded clamp attachment
Catalog # METOASK16S
Supplier:   Metro International
Grounding cables, Super Erecta®
Catalog # METOASK16S
Grounding cables, Super Erecta®
Grounding cables, Super Erecta®
Grounding cable with spring-loaded clamp attachment
Price
399,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
These grounding cables provide an effective method of electrostatic discharge for any Metro Super Erecta unit (stationary or mobile) when used in conjunction with aluminum split sleeves or ESD kit and conductive casters.
  • Cables ensure a proper path to ground when used with ESD flooring
  • Grounding cable with universal bracket fits wire and solid shelves, as well as open-wire frames
Specifications
  • Description:
    Grounding cable with spring-loaded clamp attachment
  • Seq:
    01