Baicalein ≥97%
Glass bottle for solids, 500 mg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 97% CAS No.: 491-67-8 Molecular Formula: C15H10O5 Formula Weight: 270.24 Melting Point: 268-270° Beilstein Reference: 272683 Merck Reference: 14,942 MDL No.: MFCD00017459
Synonyms: 5,6,7-Trihydroxyflavone, 5,6,7-Trihydroxy-2-phenyl-4H-chromen-4-one, 5,6,7-Trihydroxy-2-phenylchromen-4-one
- CAS Number:491-67-8
- Formula:C₁₅H₁₀O₅
- MDL Number:MFCD00017459
- Molecular Weight:270.24 g/mol
- Merck Index:14,942
- Melting Pt:268…270 °C
- Storage Temperature:Ambient
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



