Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Baicalein ≥97%

Baicalein ≥97%

Glass bottle for solids, 100 mg
Catalog # L14185.MC
Irritant
Baicalein ≥97%
Catalog # L14185.MC
CAS Number:  491-67-8
Baicalein ≥97%
Baicalein ≥97%

Glass bottle for solids, 100 mg

CAS Number:  491-67-8
Price
526,00 kr
Log in for availability
1 * 100 mg
Product Details & Documents

Purity: 97% CAS No.: 491-67-8 Molecular Formula: C15H10O5 Formula Weight: 270.24 Melting Point: 268-270° Beilstein Reference: 272683 Merck Reference: 14,942 MDL No.: MFCD00017459

Synonyms:  5,6,7-Trihydroxyflavone, 5,6,7-Trihydroxy-2-phenyl-4H-chromen-4-one, 5,6,7-Trihydroxy-2-phenylchromen-4-one
  • CAS Number:491-67-8
  • Formula:C₁₅H₁₀O₅
  • MDL Number:MFCD00017459
  • Molecular Weight:270.24 g/mol
  • Merck Index:14,942
  • Melting Pt:268…270 °C
  • Storage Temperature:Stue temperatur
Specifications

Specifications

  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.