Bisacrylamide ≥97%
500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 97% CAS No.: 110-26-9 Molecular Formula: C7H10N2O2 Formula Weight: 154.17 Storage Temperature: 2°C to 8°C Melting Point: 300° Beilstein Reference: 1706297 MDL No.: MFCD00008625
Synonyms: Bis (N,N'-methylenebisacrylamide), N,N-Methylenebisacrylamide, N,N'-Methylenediacrylamide, N,N’-Methylenebisacrylamide, N,N'-Methylenebisacrylamide, BIS
- CAS Number:110-26-9
- Formula:(H₂C=CHCONH)₂CH₂
- MDL Number:MFCD00008625
- Molecular Weight:154.17 g/mol
- Boiling Pt:125 °C (3.33 hPa)
- Melting Pt:>300 °C
- Density:1.235
- Flash Pt:138 °C
- Storage Temperature:Refrigerator
- EINECS:203-750-9
Documents
Organic Building Blocks and Intermediates
Application Notes
Specifications
Specifications
- Pk:500 g
Specifications
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



