Bisacrylamide ≥97%
100 g
Supplier: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Purity: 97% CAS No.: 110-26-9 Molecular Formula: C7H10N2O2 Formula Weight: 154.17 Storage Temperature: 2°C to 8°C Melting Point: 300° Beilstein Reference: 1706297 MDL No.: MFCD00008625
Synonymer: Bis (N,N'-methylenebisacrylamide), N,N-Methylenebisacrylamide, N,N'-Methylenediacrylamide, N,N’-Methylenebisacrylamide, N,N'-Methylenebisacrylamide, BIS
- CAS nummer:110-26-9
- Formel:(H₂C=CHCONH)₂CH₂
- MDL nummer:MFCD00008625
- Molecular Weight:154.17 g/mol
- Kogende Pt:125 °C (3.33 hPa)
- Smeltende Pt:>300 °C
- Tæthed:1.235
- Flash Pt:138 °C
- Opbevaringstemperatur:Refrigerator
- EINECS:203-750-9
Documents
Organic Building Blocks and Intermediates
Application Notes
Specifikationer
Specifikationer
- Pk:100 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



