Order Entry
Denmark
Orders LinkContactUsLinkComponent
pH indicator, bromophenol blue tablets

pH indicator, bromophenol blue tablets

Bromophenol blue tablets, pH indicator, 250 Tab.
Katalog # 162623A
Leverandør:   Lovibond
pH indicator, bromophenol blue tablets
Katalog # 162623A
Leverandør:  Lovibond
CAS nummer:  
pH indicator, bromophenol blue tablets
pH indicator, bromophenol blue tablets

Bromophenol blue tablets, pH indicator, 250 Tab.

CAS nummer:  
Price
967,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents

Indicator for pH range 2,8 to 4,4.

Can be used for measuring pH in conjunction with the Comparator 2000+ system and pH disc 2/1B

Specifications

Specifications

  • Pk:
    250 Tab.
  • Description:
    Bromophenol blue tablets, pH indicator

Specifications