Order Entry
Denmark
Orders LinkContactUsLinkComponent
VWR® Silicone Fluids

VWR® Silicone Fluids

Silicone fluid 350 cS, −40...+200 °C, 350 mm²/s, Clear, Plastic bottle, 5 kg
Catalog # 630096G
Supplier:   VWR Chemicals
VWR® Silicone Fluids
Catalog # 630096G
Supplier:  VWR Chemicals
CAS Number:  
VWR® Silicone Fluids
VWR® Silicone Fluids

Silicone fluid 350 cS, −40...+200 °C, 350 mm²/s, Clear, Plastic bottle, 5 kg

CAS Number:  
Price
14.330,00 kr
Log in for availability
1 * 5 kg
Product Details & Documents

A range of silicone fluids recommended for use in baths. Low in toxicity, inert and long lasting due to their low volatility.

  • Non corrosive
  • High flash point
  • Long service life
  • Low evaporation

Fluids possess excellent heat transfer characteristics and cover a large temperature range from −40 to +200 °C.

1 cS = 1 mm²/s

Packaging: Packed in a plastic bottle/plastic container.

Caution: Harmful vapours may be given off at elevated temperatures. It is advisable to use this material in a fume cupboard at temperatures greater than 140 °C.

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 kg
  • Colour:
    Clear
  • Description:
    Silicone fluid 350 cS
  • Temperature range:
    −40...+200 °C
  • Viscosity:
    350 mm²/s

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.