Order Entry
Denmark
Orders LinkContactUsLinkComponent
Ion exchangers

Ion exchangers

Weak acid cation exchanger, Amberlite® IRA-67, OH-, exchange capacity >1,5 mval/ml, 500 g
Catalog # 1.15959.0500
Supplier:   Merck
Ion exchangers
Catalog # 1.15959.0500
Supplier:  Merck
CAS Number:  
Ion exchangers

Weak acid cation exchanger, Amberlite® IRA-67, OH-, exchange capacity >1,5 mval/ml, 500 g

CAS Number:  
Price
2.740,00 kr
Log in for availability
1 * 500 g
Product Details & Documents

In general, ion exchangers consist of a cross-linked polymer matrix with a uniform distribution of fixed ionic groups sited throughout the resin structure. These must be balanced by a similar number of ions of the opposite charge, (counter ions) to maintain electrical neutrality. Cation exchangers therefore, exchange and enrich only cations, anion exchangers only anions. In contrast adsorber resins have a non ionic, but depending on the structure, a somewhat polar character, and don't adsorb stoichiometrically both: anions, cations, as well as uncharged compounds.

Possible applications are:
Trace enrichment by using chelating ion exchangers
Determination of total salt content of solutions and water by H+ exchange
Removals of interfering cations or anions
Chromatographic separation
Digestion of insoluble compounds
Application as a catalyst

Specifications

Specifications

  • Pk:
    500 g
  • Description:
    Weak acid cation exchanger, Amberlite® IRA-67, OH<sup>-</sup>, exchange capacity >1,5 mval/ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.