Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

2-Methylpentane ≥99%

Glass bottle, 500 mL
Catalog # L03001.AP
Flammable
Health
Irritant
Environment
2-Methylpentane ≥99%
Catalog # L03001.AP
CAS Number:  107-83-5
2-Methylpentane ≥99%
2-Methylpentane ≥99%

Glass bottle, 500 mL

CAS Number:  107-83-5
Price
2.870,00 kr
Log in for availability
1 * 500 mL
Product Details & Documents

Purity: 99+% CAS No.: 107-83-5 Molecular Formula: C6H14 Formula Weight: 86.18 Flash Point: -40°(-40°F) Refractive Index: 1.3710 Odour: Petroleum-like Melting Point: -154° Boiling Point: 61-62° Beilstein Reference: 1730735 UN No.: 1208 MDL No.: MFCD00009406

Synonyms:  1,1-Dimethylbutane, Isohexane
  • CAS Number:107-83-5
  • Formula:C₆H₁₄
  • MDL Number:MFCD00009406
  • Molecular Weight:86.18 g/mol
  • Boiling Pt:61…62 °C
  • Melting Pt:–154 °C
  • Density:0.653
  • Flash Pt:–40 °C (–40 °F)
  • Storage Temperature:Ambient
  • EINECS:203-523-4
  • UN:1208
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications