Order Entry
Denmark
Orders LinkContactUsLinkComponent
2-Methylpentane ≥99%

2-Methylpentane ≥99%

Glass bottle, 100 mL
Catalog # L03001.AE
Flammable
Health
Irritant
Environment
2-Methylpentane ≥99%
Catalog # L03001.AE
CAS Number:  107-83-5
2-Methylpentane ≥99%
2-Methylpentane ≥99%

Glass bottle, 100 mL

CAS Number:  107-83-5
Price
772,00 kr
Log in for availability
1 * 100 mL
Product Details & Documents

Purity: 99+% CAS No.: 107-83-5 Molecular Formula: C6H14 Formula Weight: 86.18 Flash Point: -40°(-40°F) Refractive Index: 1.3710 Odour: Petroleum-like Melting Point: -154° Boiling Point: 61-62° Beilstein Reference: 1730735 UN No.: 1208 MDL No.: MFCD00009406

Synonyms:  1,1-Dimethylbutane, Isohexane
  • CAS Number:107-83-5
  • Formula:C₆H₁₄
  • MDL Number:MFCD00009406
  • Molecular Weight:86.18 g/mol
  • Boiling Pt:61…62 °C
  • Melting Pt:–154 °C
  • Density:0.653
  • Flash Pt:–40 °C (–40 °F)
  • Storage Temperature:Ambient
  • EINECS:203-523-4
  • UN:1208
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.