Order Entry
Denmark
ContactUsLinkComponent
CO2 Incubator Air Jacketed with InCuSafe Stainless Steel-Copper Alloy 195 litre internal capacity (6.9 cu.ft) with 5 height adjustab le shelves. Microprocessor control with full alarm system. External Dimensions: 770 x 708 x 900mm (wdh). Internal Di 1 * 1 items
Catalog # 390-2007
Supplier:  PHC Corporation
CO2 Incubator Air Jacketed with InCuSafe Stainless Steel-Copper Alloy 195 litre internal capacity (6.9 cu.ft) with 5 height adjustab le shelves. Microprocessor control with full alarm system. External Dimensions: 770 x 708 x 900mm (wdh). Internal Di 1 * 1 items
Catalog # 390-2007
Supplier:  PHC Corporation
Frequently Bought Together