Order Entry
Denmark
Orders LinkContactUsLinkComponent
ISE sample conditioning buffers and solutions

ISE sample conditioning buffers and solutions

TISAB/NO₃, total ionic strength adjustment buffer for nitrate, -, Bottle, 4
Catalog # 662-0375
Supplier:   WTW
ISE sample conditioning buffers and solutions
Catalog # 662-0375
Supplier:  WTW
CAS Number:  
ISE sample conditioning buffers and solutions

TISAB/NO₃, total ionic strength adjustment buffer for nitrate, -, Bottle, 4

CAS Number:  
Price
3.420,00 kr
Log in for availability
1 * 4 Items
Product Details & Documents

Ion strength adjustment solutions have multiple tasks: TISAB for fluoride measurements does not only adjust the pH value, by also decomplexes the fluoride-containing complex compounds. Other ion strength adjustment solutions ensure an adapted pH value as well as the addition of a suitable conducting salt to correctly capture the concentration.

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    4
  • Description:
    TISAB/NO₃, total ionic strength adjustment buffer for nitrate
  • pH:

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.