Order Entry
Denmark
Orders LinkContactUsLinkComponent
ISE sample conditioning buffers and solutions

ISE sample conditioning buffers and solutions

ISA/Ca, ionic strength adjustment solution for calcium ions, -, Bottle, 250 mL
Catalog # WTWA140120
Supplier:   WTW
ISE sample conditioning buffers and solutions
Catalog # WTWA140120
Supplier:  WTW
CAS Number:  
ISE sample conditioning buffers and solutions

ISA/Ca, ionic strength adjustment solution for calcium ions, -, Bottle, 250 mL

CAS Number:  
Price
1.570,00 kr
Log in for availability
1 * 250 mL
Product Details & Documents

Ion strength adjustment solutions have multiple tasks: TISAB for fluoride measurements does not only adjust the pH value, by also decomplexes the fluoride-containing complex compounds. Other ion strength adjustment solutions ensure an adapted pH value as well as the addition of a suitable conducting salt to correctly capture the concentration.

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mL
  • Description:
    ISA/Ca, ionic strength adjustment solution for calcium ions
  • pH:

Specifications