Order Entry
Denmark
Orders LinkContactUsLinkComponent
ISE sample conditioning buffers and solutions

ISE sample conditioning buffers and solutions

TISAB buffer solution (F800), 1 L bottle, -, Bottle, 4
Catalog # 662-0359
Supplier:   WTW
ISE sample conditioning buffers and solutions
Catalog # 662-0359
Supplier:  WTW
CAS Number:  
ISE sample conditioning buffers and solutions

TISAB buffer solution (F800), 1 L bottle, -, Bottle, 4

CAS Number:  
Price
3.130,00 kr
Log in for availability
1 * 4 Items
Product Details & Documents

Ion strength adjustment solutions have multiple tasks: TISAB for fluoride measurements does not only adjust the pH value, by also decomplexes the fluoride-containing complex compounds. Other ion strength adjustment solutions ensure an adapted pH value as well as the addition of a suitable conducting salt to correctly capture the concentration.

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    4
  • Description:
    TISAB buffer solution (F800), 1 L bottle
  • pH:

Specifications