Order Entry
Denmark
Orders LinkContactUsLinkComponent
Syringes

Syringes

Syringe, glass, metal centre nozzle, Luer slip, grad. 0,2 ml, 2 ml
Catalog # 613-4514
Supplier:   MURRAY S
Syringes
Catalog # 613-4514
Supplier:  MURRAY S
Syringes
Syringes

Syringe, glass, metal centre nozzle, Luer slip, grad. 0,2 ml, 2 ml

Price
153,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Glass syringes with metal centre nozzle, without needles.

  • Centre nozzle with Luer slip fitting
Specifications
  • Volume:
    2 ml
  • Description:
    Syringe, glass, metal centre nozzle, Luer slip, grad. 0,2 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.