Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Rotary evaporating flasks

250 ml, 24/29 NS, 148 mm
  201-8402
 :   SciLabware
Rotary evaporating flasks
  201-8402
Rotary evaporating flasks
Rotary evaporating flasks

250 ml, 24/29 NS, 148 mm

 
1.990,00 kr
   
1 * 5 Items
 

PYREX®, borosilicate glass.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes

Ideal for use with rotary evaporators. Large surface area for rapid evaporation. Robust heavy wall construction for added strength. Florentine shape allows easy removal of solid residues.

 
  • Socket size:
    24/29 NS
  • Capacity:
    250 ml
  • Seq:
    04
  • Total height:
    148 mm