Order Entry
Denmark
Orders LinkContactUsLinkComponent
Apigenin ≥97%
Glass bottle for solids, 100 mg
  L15041.MC
Apigenin ≥97%
  L15041.MC
 :  520-36-5
Apigenin ≥97%
Apigenin ≥97%
Glass bottle for solids, 100 mg
 :  520-36-5
2.220,00 kr
1 * 100 mg
Purity: 97% CAS No.: 520-36-5 Molecular Formula: C15H10O5 Formula Weight: 270.24 Storage Temperature: 2°C to 8°C Melting Point: ca 315° dec. Beilstein Reference: 262620 Merck Reference: 14,730 MDL No.: MFCD00006831
 :  4',5,7-Trihydroxyflavone, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4-benzopyrone, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 4',6'-Dihydroxy-2-(4-hydroxyphenyl)benzo(e)oxacyclohex-2-en-4-on, 5,7-Dihydroxy-2-(4-hydroxyphenyl)chromen-4-one
  •  :520-36-5
  •  :C₁₅H₁₀O₅
  •  :MFCD00006831
  •  :270.24 g/mol
  •  :14,730
  •  :∼ 315 °C (dec.)
  •  :208-292-3
  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg