Order Entry
Denmark
Orders LinkContactUsLinkComponent
Apigenin ≥97%

Apigenin ≥97%

Glass bottle for solids, 25 mg
Catalog # L15041.MB
Irritant
Apigenin ≥97%
Catalog # L15041.MB
CAS Number:  520-36-5
Apigenin ≥97%
Apigenin ≥97%

Glass bottle for solids, 25 mg

CAS Number:  520-36-5
Price
848,00 kr
Log in for availability
1 * 25 mg
Product Details & Documents

Purity: 97% CAS No.: 520-36-5 Molecular Formula: C15H10O5 Formula Weight: 270.24 Storage Temperature: 2°C to 8°C Melting Point: ca 315° dec. Beilstein Reference: 262620 Merck Reference: 14,730 MDL No.: MFCD00006831

Synonyms:  4',5,7-Trihydroxyflavone, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4-benzopyrone, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 4',6'-Dihydroxy-2-(4-hydroxyphenyl)benzo(e)oxacyclohex-2-en-4-on, 5,7-Dihydroxy-2-(4-hydroxyphenyl)chromen-4-one
  • CAS Number:520-36-5
  • Formula:C₁₅H₁₀O₅
  • MDL Number:MFCD00006831
  • Molecular Weight:270.24 g/mol
  • Merck Index:14,730
  • Melting Pt:∼ 315 °C (dec.)
  • EINECS:208-292-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.