Order Entry
Denmark
Orders LinkContactUsLinkComponent
Abietic acid ≥75%, tech.

Abietic acid ≥75%, tech.

Glass bottle for solids, 100 g
Catalog # L13770.22
Irritant
Abietic acid ≥75%, tech.
Catalog # L13770.22
CAS Number:  514-10-3
Abietic acid ≥75%, tech.
Abietic acid ≥75%, tech.

Glass bottle for solids, 100 g

CAS Number:  514-10-3
Price
1.360,00 kr
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 75% CAS No.: 514-10-3 Molecular Formula: C20H30O2 Formula Weight: 302.45 Storage Temperature: 2°C to 8°C Odour: Characteristic Beilstein Reference: 2221451 Merck Reference: 14,7 MDL No.: MFCD03423567

  • CAS Number:514-10-3
  • Formula:C₂₀H₃₀O₂
  • MDL Number:MFCD03423567
  • Molecular Weight:302.45 g/mol
  • Merck Index:14,7
  • Storage Temperature:Refrigerator
  • EINECS:208-178-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.