Order Entry
Denmark
Orders LinkContactUsLinkComponent
2-(2-Ethoxyethoxy)ethyl acetate ≥99%

2-(2-Ethoxyethoxy)ethyl acetate ≥99%

Glass bottle, 500 mL
Catalog # L13446.AP
Irritant
2-(2-Ethoxyethoxy)ethyl acetate ≥99%
Catalog # L13446.AP
CAS Number:  112-15-2
2-(2-Ethoxyethoxy)ethyl acetate ≥99%
2-(2-Ethoxyethoxy)ethyl acetate ≥99%

Glass bottle, 500 mL

CAS Number:  112-15-2
Price
796,00 kr
Log in for availability
1 * 500 mL
Product Details & Documents

Purity: 99% CAS No.: 112-15-2 Molecular Formula: CH3CO2(CH2CH2O)2C2H5 Formula Weight: 176.21 Flash Point: 95°(203°F) Refractive Index: 1.4210 Melting Point: -25° Boiling Point: 218-219° Beilstein Reference: 1764643 Merck Reference: 14,1800 MDL No.: MFCD00041928

Synonyms:  Diethylene glycol monoethyl ether acetate
  • CAS Number:112-15-2
  • Formula:C₈H₁₆O₄
  • MDL Number:MFCD00041928
  • Molecular Weight:176.21 g/mol
  • Merck Index:14,1800
  • Boiling Pt:218…219 °C
  • Melting Pt:–25 °C
  • Density:1.012
  • Flash Pt:95 °C (203 °F)
  • Storage Temperature:Ambient
  • EINECS:203-940-1
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications