Order Entry
Denmark
Orders LinkContactUsLinkComponent
n-Hexane, anhydrous

n-Hexane, anhydrous

Glass bottle, 250 mL
Catalog # 43263.AK
Flammable
Health
Irritant
Environment
n-Hexane, anhydrous
Catalog # 43263.AK
CAS Number:  110-54-3
n-Hexane, anhydrous
n-Hexane, anhydrous

Glass bottle, 250 mL

CAS Number:  110-54-3
Price
760,00 kr
Log in for availability
1 * 250 mL
Product Details & Documents

CAS No.: 110-54-3 Molecular Formula: C6H14 Formula Weight: 86.18 Flash Point: -23°(-9°F) Physical Form: Liquid Density: 0.659 Refractive Index: 1.3750 Appearance: Colourless Odour: Weak, characteristic Melting Point: -95° Boiling Point: 69° Beilstein Reference: 1730733 Merck Reference: 14,4694 UN No.: 1208 MDL No.: MFCD00009520

Solvent in Sol-Gel precursor applications

Synonyms:  Hexyl hydride, Hexanen, Hexane
  • CAS Number:110-54-3
  • Formula:H₃C(CH₂)₄CH₃
  • MDL Number:MFCD00009520
  • Molecular Weight:86.18 g/mol
  • Merck Index:14,4694
  • Boiling Pt:69 °C
  • Melting Pt:–95 °C
  • Density:0.659
  • Flash Pt:–23 °C (–9 °F)
  • Storage Temperature:Ambient
  • EINECS:203-777-6
  • UN:1208
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL

Specifications