Order Entry
Denmark
Orders LinkContactUsLinkComponent
(±)-2-Methyltetrahydrofuran ≥99% stabilised
(±)-2-Methyltetrahydrofuran ≥99% stabilised
Glass bottle, 500 mL
  L07356.AP
(±)-2-Methyltetrahydrofuran ≥99% stabilised
  L07356.AP
 :  96-47-9
(±)-2-Methyltetrahydrofuran ≥99% stabilised
(±)-2-Methyltetrahydrofuran ≥99% stabilised
Glass bottle, 500 mL
 :  96-47-9
1.030,00 kr
1 * 500 mL
Stabilized with ca 150-400 ppm BHT
Purity: 99% CAS No.: 96-47-9 Molecular Formula: C5H10O Formula Weight: 86.13 Flash Point: -11°(12°F) Refractive Index: 1.4060 Melting Point: -136° Boiling Point: 80-82° Beilstein Reference: 102448 UN No.: 2536 MDL No.: MFCD00005367
  •  :96-47-9
  •  :C₅H₁₀O
  •  :MFCD00005367
  •  :86.13 g/mol
  •  :80…82 °C
  •  :–136 °C
  •  :0.856
  •  :–11 °C (–12 °F)
  •  :Ambient
  •  :202-507-4
  •  :3,II
  •  :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 150-400 ppm
  •  :2536
  • Pack type:
    Glass bottle
  • Pk:
    500 mL