Bromphenol blå in aqueous solution
Plastic bottle, 500 mL
Catalog # LOVI451623
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
BROMPHENOLBLÅ OPLØSNING 1 * 250 mL Catalog #LOVI451622
Product LOVI451622 is a replacement for LOVI451623 but varies by Brand, Packaging or Quantity.
Product Details & Documents
Synonyms: Bromphenol Blue Sultone Form, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein
- CAS Number:115-39-9
- Formula:C₁₉H₁₀Br₄O₅S
- MDL Number:MFCD00005875
- Molecular Weight:669.97 g/mol
- Storage Temperature:Stue temperatur
- EINECS:204-086-2
Specifications
Specifications
- Pakning:Plastic bottle
- Pakn:500 mL



