Order Entry
Denmark
Orders LinkContactUsLinkComponent
Bromophenol blue in aqueous solution

Bromophenol blue in aqueous solution

Plastic bottle, 100 mL
Catalog # LOVI451621
Supplier:   Lovibond
Bromophenol blue in aqueous solution
Catalog # LOVI451621
Supplier:  Lovibond
CAS Number:  115-39-9
Bromophenol blue in aqueous solution

Plastic bottle, 100 mL

CAS Number:  115-39-9
Price
411,00 kr
Log in for availability
1 * 100 mL
Product Details & Documents
Synonyms:  Bromphenol Blue Sultone Form, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein
  • CAS Number:115-39-9
  • Formula:C₁₉H₁₀Br₄O₅S
  • MDL Number:MFCD00005875
  • Molecular Weight:669.97 g/mol
  • Storage Temperature:Ambient
  • EINECS:204-086-2
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.