Order Entry
Denmark
Orders LinkContactUsLinkComponent
Counting chambers, Thoma
Counting chambers, Thoma
Counting chamber, dark lines, with clips, Thoma
Catalog # 720-1898
Supplier:   MARIENFELD
Counting chambers, Thoma
Catalog # 720-1898
Supplier:  MARIENFELD
Counting chambers, Thoma
Counting chambers, Thoma
Counting chamber, dark lines, with clips, Thoma
Price
671,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Designed for the determination of the yeast cell number and IVD applications.
Certifications: IVD directive 98/79/EC.
Specifications
  • Description:
    Counting chamber, dark lines, with clips, Thoma
  • Seq:
    1