Order Entry
Denmark
Orders LinkContactUsLinkComponent
Counting chambers, Bürker
Counting chambers, Bürker
Cell counting chamber, bright lined, with clamps, Bürker-Türk, 0,10 mm
Catalog # 630-1545
Supplier:   MARIENFELD
Counting chambers, Bürker
Catalog # 630-1545
Supplier:  MARIENFELD
Counting chambers, Bürker
Counting chambers, Bürker
Cell counting chamber, bright lined, with clamps, Bürker-Türk, 0,10 mm
Price
1.110,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Glass counting chambers equipped with two sets of grids, engraved into two ground, and polished surfaces on the central ridge. This central ridge is located between two elevated ridges, that are also ground and polished.
Designed for investigations of cell suspensions.
Conform to DIN 12847 and the German Calibration Ordinance
Certifications: DIN 12847 and the German Calibration Ordinance.
Delivery information: Supplied in a transparent box.
Documents
Specifications
  • Depth of chamber:
    0,10 mm
  • Description:
    Cell counting chamber, bright lined, with clamps, Bürker-Türk
  • Seq:
    05