Bensulfuron-methyl certified 1 * 0,1 g
Catalog # EHERC10498000
Leverandør: EHRENSTORFER
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Leverandør: VWR Chemicals
BUFFER PH 7,00 DOSING BOTTLE DURACAL 1 * 500 mL
Leverandør: HAMILTON BONADUZ
Potassium chloride 3 mol/l (3 N) in aqueous solution, Supelco®
Leverandør: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Leverandør: Merck
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Leverandør: VWR Chemicals
Buffer solutions in sachets
Leverandør: Mettler - Toledo
Buffer solutions in sachets, Certipur®, Supelco®
Leverandør: Merck
VWR®, Conductivity standard solutions
Leverandør: VWR Chemicals
pH buffers, DuraCal
Leverandør: HAMILTON BONADUZ
pH Standard Solutions, IUPAC, Certified
Leverandør: Hach
Ammonium acetate, Supelco®
Leverandør: Merck