Order Entry
Denmark
Orders LinkContactUsLinkComponent
1-Hexadecanol

1-Hexadecanol

Glass bottle for solids, 500 g
Catalog # ICNA0210130990
Supplier:   MP Biomedicals
1-Hexadecanol
Catalog # ICNA0210130990
Supplier:  MP Biomedicals
CAS Number:  36653-82-4
1-Hexadecanol

Glass bottle for solids, 500 g

CAS Number:  36653-82-4
Price
738,00 kr
Log in for availability
1 * 500 g
Product Details & Documents

Cetyl alcohol is used in the cosmetic industry as an opacifier in shampoos, or as an emollient, emulsifier or thickening agent in the manufacture of skin creams and lotions. It is also employed as a lubricant for nuts and bolts.

Cetyl alcohol is used in the cosmetic industry as an opacifier in shampoos, or as an emollient, emulsifier or thickening agent in the manufacture of skin creams and lotions. It is also employed as a lubricant for nuts and bolts.

Easily soluble in diethyl ether, soluble in acetone, chloroform; slightly soluble in alcohol.

Store at +4 °C

Synonyms:  Cetyl alcohol, Palmityl alcohol
  • CAS Number:36653-82-4
  • Formula:H₃C(CH₂)₁₄CH₂OH
  • MDL Number:MFCD00004760
  • Molecular Weight:242.45 g/mol
  • Merck Index:13,02037
  • Boiling Pt:305…330 °C (1013 hPa)
  • Melting Pt:46…52 °C
  • Density:0.81 g/cm³ (20 °C)
  • Flash Pt:>110 °C
  • Storage Temperature:Ambient
  • EINECS:253-149-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.