Capillary and fitting kits
Rapid resolution HT capillary kit
Catalog # HEWL5065-9947
Supplier: AGILENT HEWLETT PACKARD
Jump to:
Product Details & Documents
These starter kits contain the most widely used capillary tubing, Swagelok connectors, and fittings in a variety of sizes.
- Designed-in for highest chromatographic performance
- High quality products for lowest instrument downtime and best total cost of ownership
- OEM products fit tightest Agilent specifications for validated environments
We’ve also included our flexible stainless steel capillaries to help you make the best LC connections, regardless of equipment brand. Plus, as a special bonus, all multi-use kits (HEWL5065-9938, HEWL5065-9937, HEWL5065-9939) feature a FREE cybertool that puts more than 30 lab essentials at fingertips. For high-efficiency columns, it’s best to use narrow-diameter red tubing (Int.Ø 0,12 mm), instead of conventional green (Int.Ø 0,17 mm) tubing.
Specifications
- Description:Rapid resolution HT capillary kit
- Seq:02