Order Entry
Denmark
Orders LinkContactUsLinkComponent
Agarose

Agarose

Glass bottle for solids, 100 g
Catalog # SERA11382.02
Marketsource
Supplier:   Serva
Agarose
Catalog # SERA11382.02
Supplier:  Serva
Marketsource
CAS Number:  9012-36-6
Agarose
Agarose

Glass bottle for solids, 100 g

CAS Number:  9012-36-6
Price
Spørg for pris
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents
  • CAS nummer:9012-36-6
  • MDL nummer:MFCD00081294
  • Smeltende Pt:60…90 °C
  • Opbevaringstemperatur:Ambient
  • EINECS:232-731-8
Specifikationer

Specifikationer

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifikationer