Order Entry
Denmark
Orders LinkContactUsLinkComponent
Single-component explosives solutions

Single-component explosives solutions

Internal standard includes 3,4-dinitrotoluene, with 1000 µg/ml in methanol, 1 ml/ampule, 1 mL
Catalog # REST31452
Supplier:   Restek
Single-component explosives solutions
Catalog # REST31452
Supplier:  Restek
CAS Number:  
Single-component explosives solutions
Single-component explosives solutions

Internal standard includes 3,4-dinitrotoluene, with 1000 µg/ml in methanol, 1 ml/ampule, 1 mL

CAS Number:  
Price
407,00 kr
Log in for availability
1 * 1 mL
Product Details & Documents

Single-component explosives solutions supports US department of defense base closures and remediation.

  • Mixtures and singles supporting LC US EPA method 8330
  • Mixtures and singles supporting GC-ECD US EPA method 8095
  • Internal standards and surrogates to support both methods

These materials support nitroaromatic, nitramine, and nitroester analyses by GC-ECD (Method 8095). Compounds listed are explosives, manufacturing intermediates, or degradation products. Method 8095 mixtures contains the components at concentration ratios appropriate for ECD.

Specifications

Specifications

  • Pk:
    1 mL
  • Description:
    Internal standard includes 3,4-dinitrotoluene, with 1000 µg/ml in methanol, 1 ml/ampule

Specifications