Reference standards VOA
Dibromomethane, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule, 1
Catalog # REST30430
Supplier: Restek
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:1
- Description:Dibromomethane, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- Cat. No.:REST30430
- Formula:CH₂Br₂
- Description:Dibromomethane
- CAS No.:74-95-3
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



