Reference standards VOA
p-xylene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule, 1
Catalog # REST30426
Supplier: Restek
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:1
- Description:p-xylene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- Cat. No.:REST30426
- Formula:C₈H₁₀
- Description:p-xylene
- CAS No.:106-42-3



