Reference standards VOA
Toluene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule, 1 mL
REST30415
: Restek
- Pk:1 mL
- Description:Toluene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- Cat. No.:REST30415
- Formula:C₇H₈
- Description:Toluene
- CAS No.:108-88-3
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



