Order Entry
Denmark
Orders LinkContactUsLinkComponent
Reference standards VOA

Reference standards VOA

Pentachloroethane, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule, 1 mL
Catalog # REST30404
Supplier:   Restek
Reference standards VOA
Catalog # REST30404
Supplier:  Restek
CAS Number:  
Reference standards VOA

Pentachloroethane, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule, 1 mL

CAS Number:  
Price
280,00 kr
Log in for availability
1 * 1 mL
Product Details & Documents
Specifications

Specifications

  • Pk:
    1 mL
  • Description:
    Pentachloroethane, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
  • Cat. No.:
    REST30404
  • Formula:
    C₂HCl₅
  • Description:
    Pentachloroethane
  • CAS No.:
    76-01-7

Specifications