Reference standards VOA
Chloroethane, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule, 1
Catalog # REST30263
Supplier: Restek
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:1
- Description:Chloroethane, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- Cat. No.:REST30263
- Formula:C₂H₅Cl
- Description:Chloroethane
- CAS No.:75-00-3



