Reference standards VOA
Carbon disulphide, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule, 1
REST30258
: Restek
- Pk:1
- Description:Carbon disulphide, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- Cat. No.:REST30258
- Formula:CS₂
- Description:Carbon disulphide
- CAS No.:75-15-0
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



