Reference standards VOA
2-Butanone (Methyl ethyl ketone), 5000 µg/ml, purge and trap grade methanol, 1 ml/ampule, 1
Catalog # REST30254
Supplier: Restek
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:1
- Description:2-Butanone (Methyl ethyl ketone), 5000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- Cat. No.:REST30254
- Formula:C₄H₈O
- Description:2-Butanone (Methyl ethyl ketone)
- CAS No.:78-93-3
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



