Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Reference standards VOA

Bromobenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule, 1
Catalog # REST30250
Supplier:   Restek
Reference standards VOA
Catalog # REST30250
Supplier:  Restek
CAS Number:  
Reference standards VOA

Bromobenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule, 1

CAS Number:  
Price
306,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications

Specifications

  • Pk:
    1
  • Description:
    Bromobenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
  • Cat. No.:
    REST30250
  • Formula:
    C₆H₅Br
  • Description:
    Bromobenzene
  • CAS No.:
    108-86-1

Specifications