PCB congeners
PCB congener standard includes 2,4,4'-trichlorobiphenyl (BZ #28) with 10 µg/ml in isooctane, 1 ml/ampoule, 1 mL
Catalog # REST32283
Supplier: Restek
Jump to:
Product Details & Documents
Certified reference materials.
Manufactured and QC-tested in ISO-accredited labs.
Specifications
Specifications
- Pk:1 mL
- Description:PCB congener standard includes 2,4,4'-trichlorobiphenyl (BZ #28) with 10 µg/ml in isooctane, 1 ml/ampoule
- Cat. No.:REST32283
- Formula:C<sub>12</sub>H<sub>7</sub>Cl<sub>3</sub>
- Description:2,4,4'-trichlorobiphenyl (BZ #28)
- CAS No.:7012-37-5



