Order Entry
Denmark
Orders LinkContactUsLinkComponent
Multi-channel pipettes, electronic, variable volume, Transferpette® electronic
Multi-channel pipettes, electronic, variable volume, Transferpette® electronic
1 - 20 µl, 0,02 µl, ±1,0 %, ≤0,5 %, 0,1 - 20 / 0,5 - 20 / 1 - 50 / 0,5 - 10* / 1 - 20* µl, EU-adapter
Catalog # 613-0219
Supplier:   Brand
Multi-channel pipettes, electronic, variable volume, Transferpette® electronic
Catalog # 613-0219
Supplier:  Brand
Multi-channel pipettes, electronic, variable volume, Transferpette® electronic
Multi-channel pipettes, electronic, variable volume, Transferpette® electronic
1 - 20 µl, 0,02 µl, ±1,0 %, ≤0,5 %, 0,1 - 20 / 0,5 - 20 / 1 - 50 / 0,5 - 10* / 1 - 20* µl, EU-adapter
Price
9.560,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Transferpette® electronic pipettes combines the globally proven properties of mechanical piston operated air displacement pipettes by BRAND with the advantages of an electronic device.
  • The pipette shaft can be unscrewed and is entirely autoclavable (121 °C)
  • Five convenient programs: Pipetting, reverse pipetting, mixing, GEL-Electrophoresis and dispensing
  • Easy Calibration-Technique for making quick adjustments of the Transferpette® electronic, without tools
The Transferpette® electronic is intended for the pipetting of liquids within the following limitations: Temperature of both the instrument and solution should be between 15 °C to 40 °C (59 °F to 104 °F). Consult the manufacturer for use in temperatures outside of this range. Vapor pressure up to 500 mbar. Viscosity: 260 mPa s (260 cps).
Specifications
  • Volume:
    1 - 20 µl
  • With:
    EU-adapter
  • Seq:
    15
  • Increment:
    0,02 µl
  • Accuracy:
    ±1,0 %
  • Imprecision:
    ≤0,5 %
  • Recommended tips:
    0,1 - 20 / 0,5 - 20 / 1 - 50 / 0,5 - 10* / 1 - 20* µl
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Charging stand for 3 Transferpette® electronic up to 1000 µl, with EU plug
    Charging stand for 3 Transferpette® electronic up to 1000 µl, with EU plug
    613-4226
    Log in for availability
    1 * 1 Items - 2.070,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Individual stand for one Transferpette® electronic or -8/-12 electronic pipette, black/white
    Individual stand for one Transferpette® electronic or -8/-12 electronic pipette, black/white
    613-6937
    Log in for availability
    1 * 1 Items - 805,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Shelf/rack mount for one Transferpette® electronic or -8/-12 electronic pipette
    Shelf/rack mount for one Transferpette® electronic or -8/-12 electronic pipette
    613-6938
    Log in for availability
    1 * 1 Items - 163,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Filter for Transferpette® S/Transferpette® electronic (0,5 - 5 ml)
    Filter for Transferpette® S/Transferpette® electronic (0,5 - 5 ml)
    613-1684
    Log in for availability
    1 * 25 Items - 63,80 kr
Product Information
  • Catalog #
    Availability
    Price
  • Piston seal for Transferpette® electronic, 5 - 50 µl, multi-channel
    Piston seal for Transferpette® electronic, 5 - 50 µl, multi-channel
    BRND703343
    Log in for availability
    1 * 3 Items - 71,20 kr
Product Information
  • Catalog #
    Availability
    Price
  • Piston seal for Transferpette® S and Transferpette® electronic, 30 - 300 µl, multi channel
    Piston seal for Transferpette® S and Transferpette® electronic, 30 - 300 µl, multi channel
    BRND703346
    Log in for availability
    1 * 3 Items - 71,20 kr
Product Information
  • Catalog #
    Availability
    Price
  • Piston seal for Transferpette® electronic, 1 - 20 µl, multi-channel
    Piston seal for Transferpette® electronic, 1 - 20 µl, multi-channel
    BRND703341
    Log in for availability
    1 * 3 Items - 70,70 kr
Product Information
  • Catalog #
    Availability
    Price
  • Piston seal for Transferpette® S and Transferpette® electronic, 20 - 200 µl, multi channel
    Piston seal for Transferpette® S and Transferpette® electronic, 20 - 200 µl, multi channel
    BRND703345
    Log in for availability
    1 * 3 Items - 70,70 kr
Product Information
  • Catalog #
    Availability
    Price
  • O-ring for Transferpette® S + Transferpette® electronic, 0,5 - 10 and 1 - 20 µl, multi channel
    O-ring for Transferpette® S + Transferpette® electronic, 0,5 - 10 and 1 - 20 µl, multi channel
    BRND703380
    Log in for availability
    1 * 24 Items - 262,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Piston seal for Transferpette® S and Transferpette® electronic, 10 - 100 µl, multi channel
    Piston seal for Transferpette® S and Transferpette® electronic, 10 - 100 µl, multi channel
    BRND703344
    Log in for availability
    1 * 3 Items - 70,70 kr
Product Information
  • Catalog #
    Availability
    Price
  • O-ring for Transferpette® S + Transferpette® electronic, 5 - 50, 10 - 100, 20 - 200 and 30 - 300 µl, multi channel
    O-ring for Transferpette® S + Transferpette® electronic, 5 - 50, 10 - 100, 20 - 200 and 30 - 300 µl, multi channel
    BRND705618
    Log in for availability
    1 * 24 Items - 248,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Grease for pistons, for Transferpette® electronic -8/12, 50 - 1250 µl, approx. 1 g
    Grease for pistons, for Transferpette® electronic -8/12, 50 - 1250 µl, approx. 1 g
    613-7255
    Log in for availability
    1 * 1 Items - 118,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • O-ring for Transferpette® electronic, 50 - 1250 µl, multi channel
    O-ring for Transferpette® electronic, 50 - 1250 µl, multi channel
    613-7258
    Log in for availability
    1 * 24 Items - 314,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Piston shaft unit Transferpette® -8-12 electronic, 50 - 1250 µl
    Piston shaft unit Transferpette® -8-12 electronic, 50 - 1250 µl
    613-7259
    Log in for availability
    1 * 1 Items - 414,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Piston seal for Transferpette® S and Transferpette® electronic, 0,5 - 10 µl, multi channel
    Piston seal for Transferpette® S and Transferpette® electronic, 0,5 - 10 µl, multi channel
    BRND703340
    Log in for availability
    1 * 3 Items - 70,70 kr