Order Entry
Denmark
Orders LinkContactUsLinkComponent
Flasks, flat bottom with Quickfit® ground joints

Flasks, flat bottom with Quickfit® ground joints

50 ml, 150 mm, 24/29 NS
Catalog # 201-8442
Supplier:   SciLabware
Flasks, flat bottom with Quickfit® ground joints
Catalog # 201-8442
Supplier:  SciLabware
Flasks, flat bottom with Quickfit® ground joints
Flasks, flat bottom with Quickfit® ground joints

50 ml, 150 mm, 24/29 NS

Price
409,00 kr
Log in for availability
1 * 2 Items
Product Details & Documents

PYREX®, borosilicate glass, clear, with medium necks.

  • Very good chemical resistance
  • High temperature resistance
  • With longer neck than standard of flasks

Flat base allows use with hotplates. With labelling area.

Specifications
  • Socket size:
    24/29 NS
  • Capacity:
    50 ml
  • Height:
    150 mm