Order Entry
Denmark
Orders LinkContactUsLinkComponent
Joint clips for conical joints, POM, Quickfit®
Joint clips for conical joints, POM, Quickfit®
Mint green, 10/19
Katalog # 201-8185
Leverandør:   SciLabware
Joint clips for conical joints, POM, Quickfit®
Katalog # 201-8185
Leverandør:  SciLabware
Joint clips for conical joints, POM, Quickfit®
Joint clips for conical joints, POM, Quickfit®
Mint green, 10/19
Price
396,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents
POM, coloured. Polyacetal clips for retaining glassware having Quickfit® conical joints.
  • Easy to fit and remove
  • Can be used at temperatures up to +80 °C
  • Clips are colour coded for easy identification
Specifikationer
  • For NS:
    10/19
  • Colour:
    Mint green
  • Seq:
    01
Anbefalinger