Ion selective half-cell electrodes
2×10-5...1 mol/l, 2 - 13, 0...+80 °C, DX235-Cl-, 1 mol/l KNO₃, Cl-
Catalog # 662-2801
Supplier: Mettler - Toledo
Jump to:
Product Details & Documents
These ion selective half cells with solid-state membrane allow a quick and easy determination of concentration and activity of a variety of dissolved ions and gases.
- Modular and versatile ion selective half cells consisting of a universal shaft and a ion specific, exchangeable membrane
- Conform to RoHS (‘Restriction of Hazardous Substances’ in electrical and electronic equipment) and WEEE (‘Waste Electrical and Electronic Equipment’)
Bestillingsoplysninger: Membrane kits available separately.
Documents
Mettler InLab® Sensors
Product Literature
Specifications
- Concentration:2×10-5...1 mol/l
- For:Cl-
- Type:DX235-Cl-
- Electrolyte:1 mol/l KNO₃
- Temp.:0...+80 °C
- pH:2 - 13
Accessories
Product Information
- Catalog #AvailabilityPrice
- Membrane kit for DX280-Br⁻
Product Information
- Catalog #AvailabilityPrice
- Membrane kit for DX240-Ca²⁺
Product Information
- Catalog #AvailabilityPrice
- Membrane kit for DX239-K⁺
Product Information
- Catalog #AvailabilityPrice
- Membrane kit for DX262-NO₃⁻
Product Information
- Catalog #AvailabilityPrice
- Membrane kit for DX235-Cl⁻
Product Information
- Catalog #AvailabilityPrice
- Membrane kit for DX219-F⁻
Product Information
- Catalog #AvailabilityPrice
- Membrane kit for DX218-NH₄⁺



