Order Entry
Denmark
Orders LinkContactUsLinkComponent
Measuring jugs, conical

Measuring jugs, conical

2000 ml, 170 mm, 130 mm
Catalog # RSGA718.200
Supplier:   RSG
Measuring jugs, conical
Catalog # RSGA718.200
Supplier:  RSG
Measuring jugs, conical
Measuring jugs, conical

2000 ml, 170 mm, 130 mm

Price
1.810,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Stainless steel, Remanit 4301, with handle.

  • Good chemical resistance
  • Excellent resistance to heat
  • Smooth surfaces for fast, easy cleaning

Ideal for corrosive materials. Robust and durable.

Specifications
  • Ext.Ø:
    130 mm
  • Capacity:
    2000 ml
  • Height:
    170 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.