Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Kapselfiltre, Polycap™, Whatman™

Kapselfiltre, Polycap™, Whatman™

Polycap™ TC 36, PES, with filling bell, sterile, 0,8/0,2 µm, Stepped barb for 6 - 10 mm tubing
Catalog # 516-0983
Kapselfiltre, Polycap™, Whatman™
Catalog # 516-0983
Kapselfiltre, Polycap™, Whatman™
Kapselfiltre, Polycap™, Whatman™

Polycap™ TC 36, PES, with filling bell, sterile, 0,8/0,2 µm, Stepped barb for 6 - 10 mm tubing

Price
889,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

These Polycap™ TC capsule filters feature dual layer polytehersulfone (PES) membranes and provide efficient filtration for aqueous solutions.

  • 100% integrity-tested during manufacturing
  • PP housing thermally fused to eliminate surfactants and mold releasing agents that could affect analysis
  • Available in sterile and non sterile versions with a filling bell option
  • PES membranes protein adsorption characteristics: HAS 0,4 µg/cm²; Insulin 2,0 µg/cm²; gammaglobulin 1,5 µg/cm²

PES membranes are inherently hydrophilic, have low extractables, are biosafe, and have excellent flow rates. These characteristics make Polycap™ TC PES filters appropriate for a variety of critical filtration applications, including cleaning/rinsing pharmaceutical solutions, tissue culture media, and viral suspensions.

Polycap™ AS (Aqueous Solution) - recommended for filtering aqueous solutions. It combines a glass microfibre (GMF) prefilter and a nylon membrane, prolonging the life of the filter and allowing larger volumes and difficult samples to be filtered easily.

Polycap™ TF - designed for use with organic solvents and chemically aggressive solutions.

Polycap™ HD (Heavy Duty) - offers high filtration efficiency and excellent filtrate purity due to its materials and methods of manufacture.

Polycap™ SPF (Serum Prefilter) - optimised for prefiltration applications and typically used upstream of a Polycap™ AS or a Polycap™ PES capsule.

 : Manufactured according to ISO9001:2008 QMS standards

Specifications
  • Tilslutning:
    Stepped barb for 6 - 10 mm tubing
  • Porestørrelse:
    0,8/0,2 µm
  • Model:
    Polycap™ TC 36, PES, with filling bell, sterile
  • Seq:
    08
  • Filtreringsareal:
    440 cm²
  • Cat. No.:
    516-0983
  • Maks. tryk:
    4,1 bar