Order Entry
Denmark
Orders LinkContactUsLinkComponent
Accessories for heating circulators

Accessories for heating circulators

Stainless steel PT100 sensor with 3 m cable, 1200×6 mm, CF31, CF41, F25-HE, F25-HL, F25-ME
Catalog # JULA8981014
Supplier:   JULABO GmbH
Accessories for heating circulators
Catalog # JULA8981014
Supplier:  JULABO GmbH
Accessories for heating circulators
Accessories for heating circulators

Stainless steel PT100 sensor with 3 m cable, 1200×6 mm, CF31, CF41, F25-HE, F25-HL, F25-ME

Price
7.080,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Description:
    Stainless steel PT100 sensor with 3 m cable, 1200×6 mm
  • Seq:
    25
  • For:
    CF31, CF41, F25-HE, F25-HL, F25-ME
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.