Order Entry
Denmark
Orders LinkContactUsLinkComponent
Preparation forceps
Preparation forceps
Curved, fine, 130 mm
Catalog # HAMMHSC189-13
Supplier:   HAMMACHER KARL
Preparation forceps
Catalog # HAMMHSC189-13
Supplier:  HAMMACHER KARL
Preparation forceps
Preparation forceps
Curved, fine, 130 mm
Price
409,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Stainless steel.
Specifications
  • Type:
    Curved, fine
  • Length:
    130 mm
  • Seq:
    08